Learn More
Selleck Chemical LLC Lenalidomide10mg
SDP
Supplier:  Selleck Chemical LLC S102910MG
Encompass_Preferred
Lenalidomide (CC-5013) is aTNF-? secretion inhibitor withIC50 of 13 nM in PBMCs. Lenalidomide (CC-5013) is a ligand ofubiquitin E3 ligase cereblon (CRBN), and it causes selective ubiquitination and degradation of two lymphoid transcription factors, IKZF1 and IKZF3, by the CRBN-CRL4 ubiquitin ligase. Lenalidomide promotescleaved caspase-3 expression and inhibitVEGF expression and inducesapoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3055