Learn More
Medchemexpress LLC Lenampicillin hydrochloride | 80734-02-7 | 99.0% | 497.95 g/mol | C21H24ClN3O7S | 5 MG
SDP
Catalog No. 5000214785
Encompass_Preferred
Lenampicillin hydrochloride is the hydrochloride salt of lenampicillin, an orally active prodrug of ampicillin. It acts as a beta-lactam antibacterial that inhibits bacterial penicillin-binding proteins and is commonly used in research on suppurative skin and soft tissue infections.
  • Orally active prodrug enabling systemic absorption.
  • Beta-lactam mechanism inhibiting penicillin-binding proteins.
  • Improved absorption and reduced side effects compared with ampicillin.
  • Suitable for antibacterial activity and pharmacokinetic research.
  • Supplied as a high-purity research reagent for laboratory use.
Catalog No. 50-002-14785 Supplier Medchemexpress LLC Supplier No. HY1005005MG
Lenampicillin hydrochloride is the hydrochloride salt of lenampicillin, an orally active prodrug of ampicillin. It acts as a beta-lactam antibacterial that inhibits bacterial penicillin-binding proteins and is commonly used in research on suppurative skin and soft tissue infections.
  • Orally active prodrug enabling systemic absorption.
  • Beta-lactam mechanism inhibiting penicillin-binding proteins.
  • Improved absorption and reduced side effects compared with ampicillin.
  • Suitable for antibacterial activity and pharmacokinetic research.
  • Supplied as a high-purity research reagent for laboratory use.