Learn More
Medchemexpress LLC Lenampicillin hydrochloride | 80734-02-7 | MFCD01745174 | >99.0% | 497.95 | C21H24ClN3O7S | 10 MG
SDP
Catalog No. 5000214738
Encompass_Preferred
Lenampicillin hydrochloride is an orally active prodrug of ampicillin used as a research reagent. As a beta-lactam antibacterial, it exhibits improved oral absorption and reduced adverse effects compared with ampicillin and is commonly used in studies of suppurative skin and soft tissue infections.
  • Orally active prodrug of ampicillin.
  • Improved absorption and reduced side effects compared with ampicillin.
  • Beta-lactam antibacterial that inhibits penicillin-binding proteins.
  • Supplied as the hydrochloride salt for enhanced solubility.
  • Suitable for in vitro and in vivo research on suppurative infections.
  • Available in small research pack sizes.
Catalog No. 50-002-14738 Supplier Medchemexpress LLC Supplier No. HY10050010MG
Lenampicillin hydrochloride is an orally active prodrug of ampicillin used as a research reagent. As a beta-lactam antibacterial, it exhibits improved oral absorption and reduced adverse effects compared with ampicillin and is commonly used in studies of suppurative skin and soft tissue infections.
  • Orally active prodrug of ampicillin.
  • Improved absorption and reduced side effects compared with ampicillin.
  • Beta-lactam antibacterial that inhibits penicillin-binding proteins.
  • Supplied as the hydrochloride salt for enhanced solubility.
  • Suitable for in vitro and in vivo research on suppurative infections.
  • Available in small research pack sizes.