Learn More
Aviva Systems Biology Corp LENG4 Antibody - C-terminal region (ARP49811_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49811_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LENG4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LENG4. Alternate names: BB1, LRC4, LENG4, LPIAT, MBOA7, OACT7, hMBOA-7ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-60192