Learn More
Aviva Systems Biology Corp LENG4 Antibody - C-terminal region (ARP49810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LENG4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LENG4. Alternate names: BB1, LRC4, LENG4, LPIAT, MBOA7, OACT7, hMBOA-7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60191