Learn More
Selleck Chemical LLC leniolisib S8752-25MG
SDP
Catalog No. 5000024338
Encompass_Preferred
Leniolisib (CDZ 173) is a potent PI3K selective inhibitor with biochemical IC50 values of 0 244 0 424 2 23 and 0 011 M for PI3K PI3K PI3K and PI3K respectively
Catalog No. 50-000-24338 Supplier Selleck Chemical LLC Supplier No. S875225MG
Leniolisib (CDZ 173) is a potent PI3K selective inhibitor with biochemical IC50 values of 0 244 0 424 2 23 and 0 011 M for PI3K PI3K PI3K and PI3K respectively