Learn More
Medchemexpress LLC Leniolisib phosphate | 1354691-97-6 | 548.45 | C21H28F3N6O6P | 10 MG
SDP
Catalog No. 5000294885
Encompass_Preferred
Leniolisib phosphate is the phosphate salt of leniolisib, a potent and selective phosphoinositide 3-kinase delta (PI3Kδ) inhibitor supplied as a research reagent for laboratory studies into PI3Kδ signaling and related immunodeficiency disorders.
  • Potent and selective PI3Kδ inhibition for targeted signaling studies.
  • Phosphate salt form improves aqueous solubility for biological assays.
  • Characterized chemical with defined formula and molecular weight for reproducible experiments.
  • Suitable for biochemical and cellular assay development.
  • Intended for research use only.
Catalog No. 50-002-94885 Supplier Medchemexpress LLC Supplier No. HY17635A10MG
Leniolisib phosphate is the phosphate salt of leniolisib, a potent and selective phosphoinositide 3-kinase delta (PI3Kδ) inhibitor supplied as a research reagent for laboratory studies into PI3Kδ signaling and related immunodeficiency disorders.
  • Potent and selective PI3Kδ inhibition for targeted signaling studies.
  • Phosphate salt form improves aqueous solubility for biological assays.
  • Characterized chemical with defined formula and molecular weight for reproducible experiments.
  • Suitable for biochemical and cellular assay development.
  • Intended for research use only.