Learn More
Medchemexpress LLC Leniolisib phosphate | 1354691-97-6 | 99.1% | 548.45 | 5 MG
SDP
Catalog No. 5000387309
Encompass_Preferred
Leniolisib phosphate is a potent and selective PI3Kδ inhibitor. It has the potential for immunodeficiency disorders treatment.
  • Potent and selective PI3Kδ inhibitor
  • Potential for immunodeficiency disorders treatment
  • Appears as a solid, white to off-white in color
Catalog No. 50-003-87309 Supplier Medchemexpress LLC Supplier No. HY17635A5MG
Leniolisib phosphate is a potent and selective PI3Kδ inhibitor. It has the potential for immunodeficiency disorders treatment.
  • Potent and selective PI3Kδ inhibitor
  • Potential for immunodeficiency disorders treatment
  • Appears as a solid, white to off-white in color