Learn More
Medchemexpress LLC Leniolisib (phosphate) | 1354691-97-6 | 99.1% | 548.45 | 50 MG
SDP
Catalog No. 5000386962
Encompass_Preferred
Leniolisib (CDZ173) phosphate is a potent and selective PI3Kδ inhibitor. It has potential for the treatment of immunodeficiency disorders. This product is for research use only and is not sold to patients.
  • Potent and selective PI3Kδ inhibitor
  • Potential for immunodeficiency disorders treatment
  • Suppresses increased pathway activity (phosphorylation of AKT or S6) caused by APDS mutant p110δ in a concentration-dependent manner
Catalog No. 50-003-86962 Supplier Medchemexpress LLC Supplier No. HY17635A50MG
Leniolisib (CDZ173) phosphate is a potent and selective PI3Kδ inhibitor. It has potential for the treatment of immunodeficiency disorders. This product is for research use only and is not sold to patients.
  • Potent and selective PI3Kδ inhibitor
  • Potential for immunodeficiency disorders treatment
  • Suppresses increased pathway activity (phosphorylation of AKT or S6) caused by APDS mutant p110δ in a concentration-dependent manner