Learn More
Research Products International Corp LB Broth Powder [Lennox L Broth], 2 Kilograms
SDP
Catalog No. 50488758
Encompass_Preferred
For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade.
Dissolve 20 grams in 1 liter distilled water. Then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes.
Ingredients:
Casein Digest of Peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 5 g/L
Catalog No. 50-488-758 Supplier Research Products International Corp Supplier No. L240602000.0
For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade.
Dissolve 20 grams in 1 liter distilled water. Then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes.
Ingredients:
Casein Digest of Peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 5 g/L