Learn More
Research Products International Corp LB Broth, Granulated [Lennox L Broth], 500 Grams
SDP
Catalog No. 50488762
Encompass_Preferred
For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade.
Dissolve 20 grams in 1 liter distilled water. Then adjust the pH to 7.2. Heat and frequently agitate to completely dissolve. Material is specially granulated to reduce dust and increase solubility rate. Sterilize by autoclaving for 15 minutes.
Ingredients:
Tryptone:10 g/l
Sodium Chloride: 5 g/l
Yeast Extract: 5 g/l
Catalog No. 50-488-762 Supplier Research Products International Corp Supplier No. L24066500.0
For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade.
Dissolve 20 grams in 1 liter distilled water. Then adjust the pH to 7.2. Heat and frequently agitate to completely dissolve. Material is specially granulated to reduce dust and increase solubility rate. Sterilize by autoclaving for 15 minutes.
Ingredients:
Tryptone:10 g/l
Sodium Chloride: 5 g/l
Yeast Extract: 5 g/l