Learn More
Research Products International Corp LB Broth 1 Gram Buffered Capsules [Lennox L Broth], 500 Grams
SDP
Catalog No. 50550420
Encompass_Preferred
RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments, dust and smells from the lab. For propagation and maintenance of E.Coli. Low Salt Formula. Molecular Biology Grade.
Add 20 capsules per liter of water. Heat and stir until capsule is completely dissolved. Buffer is set for 7.2 at 20°C using a 10mM solution of TRIS/TRIS HCl. Sterilize by autoclaving for 15 minutes.
Ingredients:
Casein digest of peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 5 g/L
Catalog No. 50-550-420 Supplier Research Products International Corp Supplier No. L24061500.0
RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments, dust and smells from the lab. For propagation and maintenance of E.Coli. Low Salt Formula. Molecular Biology Grade.
Add 20 capsules per liter of water. Heat and stir until capsule is completely dissolved. Buffer is set for 7.2 at 20°C using a 10mM solution of TRIS/TRIS HCl. Sterilize by autoclaving for 15 minutes.
Ingredients:
Casein digest of peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 5 g/L