Learn More
Electron Microscopy Sciences EMS Residual Oil Remover Dropper style 1 OZ
SDP
Catalog No. NC1476863
Encompass_Preferred
ROR is specifically designed to emulsify and remove microscopic oil residues, a by product of environmental pollution, from the surface and pores of glass and plastic.
When you clean an optical surface with ROR you will observe a measurable difference in brightness and clarity. For the first time, you will experience pollution free, unimpeded vision. Because ROR removes microscopic oil residues, dust has nothing to adhere to allowing the surface to remain cleaner longer.
ROR is safe and versatile for all delicate surfaces. It has been tested and used on everything from multicoated lenses to beryllium surfaced contacts and has proven time and again to be the finest and safest optical cleaning treatment on the market today.
Catalog No. NC1476863 Supplier Electron Microscopy Sciences Supplier No. 7431930
ROR is specifically designed to emulsify and remove microscopic oil residues, a by product of environmental pollution, from the surface and pores of glass and plastic.
When you clean an optical surface with ROR you will observe a measurable difference in brightness and clarity. For the first time, you will experience pollution free, unimpeded vision. Because ROR removes microscopic oil residues, dust has nothing to adhere to allowing the surface to remain cleaner longer.
ROR is safe and versatile for all delicate surfaces. It has been tested and used on everything from multicoated lenses to beryllium surfaced contacts and has proven time and again to be the finest and safest optical cleaning treatment on the market today.