Learn More
Pyramex™ Hygienic Cleaning Wipes
Catalog No. 19555091 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Scratch-resistant, UV-treated
Catalog No. Lens Coating
19-555-091 Scratch-resistant, UV-treated
1 options
This item is not returnable. View return policy
Catalog No. 19-555-091 Supplier Pyramex™ Supplier No. HCW100A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hygienic Cleaning Wipes for use on respirators, personal equipment, self-contained breathing apparatus, and hard hat suspensions.
  • Removes body oils and perspiration from respirators and protective equipment
  • Wipes contain alcohol
  • 100 individually packaged pre-moistened hygienic wipes
Specifications
Height (English) 5 in.
Width (English) 8 in.
Lens Coating Scratch-resistant, UV-treated
Description Contains alcohol
Product Type Hygienic Wipes
No. per Pack 100 Pk.
Material Microfiber
Disposable Single-use
Autoclavable Not Autoclavable