Learn More
Grainger Radians Lens Cleaning Liquid, Anti-Fog, Anti-Static, 4/Pk.
Catalog No. 502786598
Encompass_Preferred
This item has to be ordered in multiples of 4 per supplier requirements.
VisionAid lens cleaning liquid is designed to effectively remove grime and oily films from optical surfaces while providing essential anti-fog and anti-static protection. This specialized solution helps ensure clarity and safety for various lens materials, making it a reliable choice for maintaining protective eyewear.
  • Safe for use on glass, plastic, and polycarbonate lenses
  • Loosens grime and oily film for easy removal
  • Cleans and coats lenses with anti-fog and anti-static solution in one step
  • Silicone-free formulation
  • OSHA compliant
  • Container features a twist-off cap for easy application
  • Slight alcoholic scent
Catalog No. 50-278-6598 Supplier Grainger Supplier No. 198EA1
This item has to be ordered in multiples of 4 per supplier requirements.
VisionAid lens cleaning liquid is designed to effectively remove grime and oily films from optical surfaces while providing essential anti-fog and anti-static protection. This specialized solution helps ensure clarity and safety for various lens materials, making it a reliable choice for maintaining protective eyewear.
  • Safe for use on glass, plastic, and polycarbonate lenses
  • Loosens grime and oily film for easy removal
  • Cleans and coats lenses with anti-fog and anti-static solution in one step
  • Silicone-free formulation
  • OSHA compliant
  • Container features a twist-off cap for easy application
  • Slight alcoholic scent