Learn More
Grainger VisionAid Lens Cleaning Liquid, Clear, Anti-Fog, Anti-Static, 16 oz, 24/pk.
Catalog No. 502788103
Encompass_Preferred
This item has to be ordered in multiples of 24 per supplier requirements.
This lens cleaning liquid is designed for the effective cleaning of eye and face protection lenses, including glass, plastic, and polycarbonate materials. The formulation features isopropyl alcohol for efficient soil removal while remaining anti-fog and anti-static for enhanced clarity and convenience in professional environments.
  • Compatible with glass, plastic, and polycarbonate lenses
  • Anti-fog properties for maintained visibility
  • Anti-static formula to reduce dust attraction
  • Liquid mixture for versatile application
Catalog No. 50-278-8103 Supplier Grainger Supplier No. 198EA0
This item has to be ordered in multiples of 24 per supplier requirements.
This lens cleaning liquid is designed for the effective cleaning of eye and face protection lenses, including glass, plastic, and polycarbonate materials. The formulation features isopropyl alcohol for efficient soil removal while remaining anti-fog and anti-static for enhanced clarity and convenience in professional environments.
  • Compatible with glass, plastic, and polycarbonate lenses
  • Anti-fog properties for maintained visibility
  • Anti-static formula to reduce dust attraction
  • Liquid mixture for versatile application