Learn More
Pyramex Safety Products Lens Cleaning Tissues
Catalog No. 19555137 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Packaging:
Box
Canister
No. per Pack:
100 Pk.
300 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging No. per Pack
19-555-137 Canister 100 Pk.
19-555-140 Box 300 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-555-137 Supplier Pyramex Safety Products Supplier No. LCC100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lens Cleaning Tissues for plastic, glass, and polycarbonate lenses.
  • Available in a canister with 100 lens cleaning tissues or a box with 300 lens cleaning tissues.
  • Safe and effective with all lens coatings.
  • Ideal for glasses, goggles, and face shields.
  • Anti-fog, anti-static formula.
Specifications
Lens Coating Anti-fog
Product Type Lens Cleaning Tissue
No. per Pack 100 Pk.
Material Microfiber
Disposable Single-use
Autoclavable Not Autoclavable
Packaging Canister