Learn More
Pyramex Safety Products Lens Cleaning Tissues
Catalog No. 19555140 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Packaging:
Box
Canister
No. per Pack:
100 Pk.
300 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging No. per Pack
19-555-140 Box 300 Pk.
19-555-137 Canister 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-555-140 Supplier Pyramex Safety Products Supplier No. LT300
Add to Cart
Edge
Add to Cart
Lens Cleaning Tissues for plastic, glass, and polycarbonate lenses.
  • Available in a canister with 100 lens cleaning tissues or a box with 300 lens cleaning tissues.
  • Safe and effective with all lens coatings.
  • Ideal for glasses, goggles, and face shields.
  • Anti-fog, anti-static formula.
Specifications
Lens Coating Scratch-resistant
Product Type Lens Cleaning Tissue
No. per Pack 300 Pk.
Material Tissue
Disposable Single-use
Autoclavable Not Autoclavable
Packaging Box