Learn More
Grainger Lens Cover 4-1/2 X 3-3/8 PK5
Catalog No. 502749207
Encompass_Preferred
The Miller Electric Front Lens Cover for the Classic Pro welding helmet is a replacement clear plastic lens cover that protects the auto-darkening filter lens. It measures 4.5 x 5.25 inches and is compatible with Classic Pro welding helmets. The lens cover easily snaps into place to shield the filter lens from sparks, spatter and debris. It should be replaced when it becomes scratched or pitted for optimal visibility. This genuine Miller part ensures continued safety and performance.
Catalog No. 50-274-9207 Supplier Grainger Supplier No. 13Z857
The Miller Electric Front Lens Cover for the Classic Pro welding helmet is a replacement clear plastic lens cover that protects the auto-darkening filter lens. It measures 4.5 x 5.25 inches and is compatible with Classic Pro welding helmets. The lens cover easily snaps into place to shield the filter lens from sparks, spatter and debris. It should be replaced when it becomes scratched or pitted for optimal visibility. This genuine Miller part ensures continued safety and performance.