Learn More
Grainger Bullard® Lens Covers, Clear, Polyester Film, 17 in. x 6 in., 25/Pk.
Catalog No. 502794382
Encompass_Preferred
These clear, polyester film lens covers are designed to protect the front of respirator lenses on compatible headgear. They maintain optimal visibility by shielding the underlying lens from wear and contaminants, and can be easily removed and replaced.
  • Compatible with CC20 series hoods
  • Constructed from polyester film
  • Designed for front lens protection
  • Dimensions of 17 in. x 6 in.
  • Packaged in a quantity of 25/Pk.
Catalog No. 50-279-4382 Supplier Grainger Supplier No. 1AAK3
These clear, polyester film lens covers are designed to protect the front of respirator lenses on compatible headgear. They maintain optimal visibility by shielding the underlying lens from wear and contaminants, and can be easily removed and replaced.
  • Compatible with CC20 series hoods
  • Constructed from polyester film
  • Designed for front lens protection
  • Dimensions of 17 in. x 6 in.
  • Packaged in a quantity of 25/Pk.