Learn More
Bioworld LENS CULINARISLENTIL) - L
Catalog No. 50198738
Encompass
LENS CULINARIS (LENTIL) - LECTIN: Lens culinaris (Lentil) Lectin isolated from lentil seeds. Specifications: Primary sugar specificity: Glucose/mannose Preferential binding activity: alpha-linked mannose (divalent cations needed)
Catalog No. 50-198-738 Supplier Bioworld Supplier No. 215100062
LENS CULINARIS (LENTIL) - LECTIN: Lens culinaris (Lentil) Lectin isolated from lentil seeds. Specifications: Primary sugar specificity: Glucose/mannose Preferential binding activity: alpha-linked mannose (divalent cations needed)