Learn More
Bioworld LENS CULINARISLENTIL) - L
Supplier:  Bioworld 215100061
Encompass_Preferred
LENS CULINARIS (LENTIL) - LECTIN: Lens culinaris (Lentil) Lectin isolated from lentil seeds. Specifications: Primary Sugar Specificity: Glucose/Mannose Preferential binding activity: alpha-linked mannose (divalent cations needed).
Catalog No. 50-198-737
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.