Learn More
Bioworld Lens culinaris (Lentil) Lectin (LCA/LCH) - Rhodamine (TRITC)
Supplier:  Bioworld 217610341
Encompass
Technical Specifications:Ligand Source:Lens culinarisMolecular Weight:50 kDaCarbohydrate Specificity:αMan (High Mannose), Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution:0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):7.6-8.4Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-199-147