Learn More
Bioworld Lens culinaris (Lentil) Lectin (LCA/LCH) - FITC
Catalog No. 50199145
Encompass
Technical Specifications:Ligand Source:Lens culinarisMolecular Weight: 50 kDaCarbohydrate Specificity:αMan (High Mannose), Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution: 0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity: N/AConjugate: FITCForm: LiquidIsoelectric (pI): 7.6-8.4Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-199-145 Supplier Bioworld Supplier No. 217610331
Add to Cart
Add to Cart
Technical Specifications:Ligand Source:Lens culinarisMolecular Weight: 50 kDaCarbohydrate Specificity:αMan (High Mannose), Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution: 0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity: N/AConjugate: FITCForm: LiquidIsoelectric (pI): 7.6-8.4Storage: -20°CApplication note: Calcium and Manganese are required for binding.