Learn More
Supertek™ Lens, Double Concave, 50mmD
GREENER_CHOICESCI_ED
Catalog No. S012326 Shop All Supertek Scientific Products
Change view
Click to view available options
Diameter (Metric):
50 mm
Catalog No. Diameter (Metric)
S012326 50 mm
1 options
Catalog No. S012326 Supplier Supertek™ Supplier No. PH30078/4
Only null left
Add to Cart
Add to Cart
Double Concave lens made of high quality glass with outstanding optical properties
  • Double concave lens.
  • Made of high quality glass with outstanding optical properties.
  • 50 mm diameter with 200 mm focal length.
Specifications
Diameter (Metric) 50 mm
Product Type Concave Lens
Shape Concave Circle