Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Lens Frame Kit, Replacement

Catalog No. 1798596F Shop All 3M Products
Change view
Click to view available options
Quantity:
5/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-985-96F 5/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-985-96F Supplier 3M™ Supplier No. 7000125978
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replacement part for 3M™ Full Facepieces 7000 Series. Use to attach 3M™ Lens 7884 to facepiece. Does not include lens.

  • Approved 3M replacement part
  • Recommended for 3M Full Facepiece 7000 Series (sold separately)
  • Use to attach 3M™ Lens 7884 to facepiece (sold separately)
  • Replacement lens frame kit can extend service life of facepiece
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Replacement Parts and Accessories
For Use With (Application) Facility Maintenance, Assembly and Mechanical, Painting, Cleaning
For Use With (Equipment) 3M™ Full Facepiece 7000 Series
Model 7885
Quantity 5/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.