Learn More
Fisherbrand™ Lens Paper
SDPSCI_ED
Catalog No. p-2381140
Change view
Click to view available options
Quantity:
100 Sheets/Book, 12 Books/Pk.
50 Sheets/Book, 12 Books/Pk.
Dimensions (L x W):
4 x 6 in.
6 x 8 in. sheets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dimensions (L x W)
S47605 50 Sheets/Book, 12 Books/Pk. 4 x 6 in.
S47606 100 Sheets/Book, 12 Books/Pk. 6 x 8 in. sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S47605 Supplier Fisherbrand™ Supplier No. 20 20 5115/EMD
Only null left
Add to Cart
Add to Cart
For cleaning glass lenses.
Booklet
  • Never scratches glass lenses, objectives, or other delicate surfaces
  • Doesn't become harsh; doesn't readily collect dust or grease
  • Virtually lint-free sheets are glued and bound in a booklet for easy removal
Specifications
Product Type Lens Cleaning Paper
Dimensions (L x W) 4 x 6 in.
For Use With (Application) For cleaning glass lenses
Quantity 50 Sheets/Book, 12 Books/Pk.