Learn More
Tiffen Company LENS PAPER 50PK/CS
Catalog No. NC9731809
Encompass_Preferred
TIFFEN Lens Cleaning Paper. Designed for use with delicate lenses and sensitive surfaces. Ideal for coated optics and laser cleaning applications. Features: High wet strength, extremely soft and non-abrasive, Meets government lens paper specifications; Type 1, Class 1 product specifications AA-50177. Sales unit = One (1) box containing 50 envelopes containing 50 sheets (2-3/4 x 4-1/2 in.) of tissue per envelope.
Catalog No. NC9731809 Supplier Tiffen Company Supplier No. EK1546027T
TIFFEN Lens Cleaning Paper. Designed for use with delicate lenses and sensitive surfaces. Ideal for coated optics and laser cleaning applications. Features: High wet strength, extremely soft and non-abrasive, Meets government lens paper specifications; Type 1, Class 1 product specifications AA-50177. Sales unit = One (1) box containing 50 envelopes containing 50 sheets (2-3/4 x 4-1/2 in.) of tissue per envelope.