Learn More
Electron Microscopy Sciences Replacement Lens Plate for #68108-06, 2.50x, 8" range
SDP
Catalog No. 5026450
Encompass_Preferred
Lightweight, adjustable headband for comfort provides close-up magnification while leaving both hands free for working. Tilt up and away when not in use. Single lens with no center post means clear, comfortable, unobstructed vision with less eye strain and fatigue. Replacement lens for Headband Magnifier #68108-06.
Magnification: 2.50x
Range: 8"
Catalog No. 50-264-50 Supplier Electron Microscopy Sciences Supplier No. 6810816
Lightweight, adjustable headband for comfort provides close-up magnification while leaving both hands free for working. Tilt up and away when not in use. Single lens with no center post means clear, comfortable, unobstructed vision with less eye strain and fatigue. Replacement lens for Headband Magnifier #68108-06.
Magnification: 2.50x
Range: 8"