Learn More
Electron Microscopy Sciences Optivison™ Head Band Magnifier Lens Plate Replacement 2.75x
SDP
Catalog No. 5030518
Encompass_Preferred
Features a dial adjustment headband that fits any head size. Wellmade magnifiers they are lightweight and durable, and fit over eyeglasses. Adjustable pivot assembly on visor can be set to desired tension, so it will stay in place when tilted. Lenses are ground and polished prismatic. Lens plate replacement is also available. Lens plate for catalog # 68107-06 head band magnifier.
Magnification: 2.75x
Catalog No. 50-305-18 Supplier Electron Microscopy Sciences Supplier No. 6810716
Features a dial adjustment headband that fits any head size. Wellmade magnifiers they are lightweight and durable, and fit over eyeglasses. Adjustable pivot assembly on visor can be set to desired tension, so it will stay in place when tilted. Lenses are ground and polished prismatic. Lens plate replacement is also available. Lens plate for catalog # 68107-06 head band magnifier.
Magnification: 2.75x