Learn More
Electron Microscopy Sciences Optivison™ Head Band Magnifier Lens Plate Replacement 3.5x
SDP
Catalog No. 5030519
Encompass_Preferred
Features a dial adjustment headband that fits any head size. Wellmade magnifiers they are lightweight and durable, and fit over eyeglasses. Adjustable pivot assembly on visor can be set to desired tension, so it will stay in place when tilted. Lenses are ground and polished prismatic. Lens plate replacement is also available. Lens plate for catalog # 68107-07 head band magnifier.
Magnification: 3.5x
Catalog No. 50-305-19 Supplier Electron Microscopy Sciences Supplier No. 6810717
Features a dial adjustment headband that fits any head size. Wellmade magnifiers they are lightweight and durable, and fit over eyeglasses. Adjustable pivot assembly on visor can be set to desired tension, so it will stay in place when tilted. Lenses are ground and polished prismatic. Lens plate replacement is also available. Lens plate for catalog # 68107-07 head band magnifier.
Magnification: 3.5x