Learn More
Electron Microscopy Sciences EMS Lens Tissue Booklet, 50 Sheets
SDP
Catalog No. 50949076
Encompass_Preferred
EMS Lens Tissue is the world's finest optical tissue. It's extremely soft and virtually lint-free. Safe for any optical surface such as camera lenses, microscopes, telescopes, and eye-glasses. Each tissue is perforated for easy removal from the booklet.
Catalog No. 50-949-076 Supplier Electron Microscopy Sciences Supplier No. 7171201
EMS Lens Tissue is the world's finest optical tissue. It's extremely soft and virtually lint-free. Safe for any optical surface such as camera lenses, microscopes, telescopes, and eye-glasses. Each tissue is perforated for easy removal from the booklet.