Learn More
Takara Bio Lenti-X™ 293T Cell Line, 1 ml
SDP
Catalog No. NC9834960
Encompass
The Lenti-X 293T Cell Line is a sub-clone of the transformed human embryonic kidney cell line, HEK 293, which is highly transfectable and supports high levels of viral protein expression. When transfected with the Lenti-X HTX Packaging System and a lentiviral vector, these cells are capable of producing lentiviral titers as high as >108 ifu/ml, as determined by flow cytometry. The cell line also constitutively expresses the simian virus 40 (SV40) large T antigen. 1ml
Catalog No. NC9834960 Supplier Takara Bio Supplier No. 632180
The Lenti-X 293T Cell Line is a sub-clone of the transformed human embryonic kidney cell line, HEK 293, which is highly transfectable and supports high levels of viral protein expression. When transfected with the Lenti-X HTX Packaging System and a lentiviral vector, these cells are capable of producing lentiviral titers as high as >108 ifu/ml, as determined by flow cytometry. The cell line also constitutively expresses the simian virus 40 (SV40) large T antigen. 1ml