Learn More
Takara Bio Lenti-X™ Packaging Single Shots (Ecotropic), 16 reactions
SDP
Supplier:  Takara Bio 631278
Encompass
Lenti-X Packaging Single Shots (Ecotropic) provide a one-step production method to generate high-titer lentivirus. Each tube of Lenti-X Packaging Single Shots (Ecotropic) contains premeasured, lyophilized Xfect Transfection Reagent and an optimized formulation of Lenti-X lentiviral packaging plasmids. High-titer ecotropic lentivirus is produced by adding the lentiviral vector of choice in sterile water to the tube, reconstituting, and then using the mixture to transfect 293T cells (e.g., Lenti-X 293T Cell Line, Cat. No. 632180) in a 10-cm dish.
Catalog No. NC1380015