Learn More
Takara Bio LENTI-X QRT-PCR TITRATION
SDP
Catalog No. NC9916222
Encompass
Retroviral and lentiviral titration kits that measure virus titers using SYBR Green-based, real-time PCR. Viral copy numbers are determined by comparing the sampleAEs Ct to a standard curve derived from a calibrated control template.
Catalog No. NC9916222 Supplier Takara Bio Supplier No. 631235
Retroviral and lentiviral titration kits that measure virus titers using SYBR Green-based, real-time PCR. Viral copy numbers are determined by comparing the sampleAEs Ct to a standard curve derived from a calibrated control template.