Learn More
Boca Scientific Inc LentiBlast Premium Transduction Enhancer, 1500ul
Catalog No. NC1747708
Encompass
LentiBlast Premium Transduction Enhancer is designed to increase lentiviral infection and transduction efficiency in a wide range of cell types, from CD34+ hematopoietic stem cells to primary cells or cell lines such as HT1080 or SH SY5Y. LentiBlast Premium is composed of only one reagent, allowing for a new single-step protocol. With LentiBlast Premium, neutralizing electrostatic repulsions between membranes and viral particles and enhancing viral fusion with cell membranes can occur simultaneously. Due to a favorable membrane permeable effect limiting the transmembrane potential changes, LentiBlast Premium is non-toxic and totally compatible with cell viability.
Catalog No. NC1747708 Supplier Boca Scientific Inc Supplier No. LBPX1500
LentiBlast Premium Transduction Enhancer is designed to increase lentiviral infection and transduction efficiency in a wide range of cell types, from CD34+ hematopoietic stem cells to primary cells or cell lines such as HT1080 or SH SY5Y. LentiBlast Premium is composed of only one reagent, allowing for a new single-step protocol. With LentiBlast Premium, neutralizing electrostatic repulsions between membranes and viral particles and enhancing viral fusion with cell membranes can occur simultaneously. Due to a favorable membrane permeable effect limiting the transmembrane potential changes, LentiBlast Premium is non-toxic and totally compatible with cell viability.