Learn More
Mayflower Bioscience LentiBOOST-P Pharma Grade Lentivirus Transduction Enhancer Solution
SDP
Catalog No. NC1801469
Encompass
LentiBOOSTTM is a highly effective, non-cytotoxic transduction enhancer for preclinical and clinical application of lentiviral vectors. As universally acting (receptor independent) adjuvant it can be applied to a wide range of clinically relevant cell types including CD34+ hematopoietic stem cells (HSCs), primary T cells and NK cells. These unique features make it a promising candidate to improve clinical transduction protocols for ex vivo gene therapies and CAR-T cell therapies.
Catalog No. NC1801469 Supplier Mayflower Bioscience Supplier No. SBPLV10111
LentiBOOSTTM is a highly effective, non-cytotoxic transduction enhancer for preclinical and clinical application of lentiviral vectors. As universally acting (receptor independent) adjuvant it can be applied to a wide range of clinically relevant cell types including CD34+ hematopoietic stem cells (HSCs), primary T cells and NK cells. These unique features make it a promising candidate to improve clinical transduction protocols for ex vivo gene therapies and CAR-T cell therapies.