Learn More
Mayflower Bioscience LentiBOOST Research Grade Lentivirus Transduction Enhancer Solution, 200 Transductions
SDP
Catalog No. NC1889820
Encompass_Preferred
LentiBOOST is a chemical transduction enhancer simultaneously neutralizing electrostatic repulsion between cell membrane and virion and enhancing viral membrane fusion. In contrast to other chemical enhancers, LentiBOOST shows no toxic effects even in high concentrations. Therefore LentiBOOST is highly recommended for hard to transduce cells even in the presence of other transduction enhancers. LentiBOOST-R Research grade material is used for basic research only and is not transferable to the clinic. This is the latest technology which has been produced by Revvity and introduced by Mayflower Bioscience. 
Catalog No. NC1889820 Supplier Mayflower Bioscience Supplier No. SBPLV10103
LentiBOOST is a chemical transduction enhancer simultaneously neutralizing electrostatic repulsion between cell membrane and virion and enhancing viral membrane fusion. In contrast to other chemical enhancers, LentiBOOST shows no toxic effects even in high concentrations. Therefore LentiBOOST is highly recommended for hard to transduce cells even in the presence of other transduction enhancers. LentiBOOST-R Research grade material is used for basic research only and is not transferable to the clinic. This is the latest technology which has been produced by Revvity and introduced by Mayflower Bioscience. 