Learn More
Cellecta Inc LentiFuge Viral Concentration Reagent, 1 ml, 1000X solution, sterile, cell culture grade, non-toxic, concentrates virus from 1 L of supernatant
SDP
Catalog No. NC1767324
Encompass
LentiFuge Viral Concentration Reagent, 1 ml, 1000X solution, sterile, cell culture grade, non-toxic, concentrates virus from 1 L of supernatant
Catalog No. NC1767324 Supplier Cellecta Inc Supplier No. LFVC1
LentiFuge Viral Concentration Reagent, 1 ml, 1000X solution, sterile, cell culture grade, non-toxic, concentrates virus from 1 L of supernatant