Learn More
OriGene Lentiviral Control Particles, x10^7 TU/ml, 5x500 ul
SDP
Catalog No. 5000600196
Encompass
Control Lenti particles, Scrambled shRNA, Expressing GFP and Puro, 1x10^7 TU/ml, 5x0.5 mlType: Lenti-shRNA Control ParticlesMammalian Cell Selection: PuromycinControl Type: NegativeFeatures: Lenti particles of Cat# TR30021 carrying a 29-mer scrambled sequence cassette and serve as a negative control.Fluorescent Label: GFPVector: pGFP-C-shLenti shRNA VectorViral Packaging: Lentiviral
Catalog No. 50-006-00196 Supplier OriGene Supplier No. TR30021V5
Control Lenti particles, Scrambled shRNA, Expressing GFP and Puro, 1x10^7 TU/ml, 5x0.5 mlType: Lenti-shRNA Control ParticlesMammalian Cell Selection: PuromycinControl Type: NegativeFeatures: Lenti particles of Cat# TR30021 carrying a 29-mer scrambled sequence cassette and serve as a negative control.Fluorescent Label: GFPVector: pGFP-C-shLenti shRNA VectorViral Packaging: Lentiviral