Learn More
OriGene Lentiviral Packaging Kits, 5x10 reactions
SDP
Catalog No. NC1749550
Encompass_Preferred
Lenti-vpak packaging kit - packaging plasmids and transfection reagent, 5 packsComponents: Five kits each containing: 1 vial of packaging plasmids (60 ug) – lyophilized 1 vial of Turbofectin (400 ul) – liquidQuality Control: The reagent components have been successfully tested in transfecting HEK293 cells and producing pseudoviral particles for transduction.Stability: Stable for 1 year from receipt of products under proper storage and handling condition.Storage: Store in 4°C refrigerator upon arrival.Type: Lentiviral Packaging Kits
Catalog No. NC1749550 Supplier OriGene Supplier No. TR30037P5
Lenti-vpak packaging kit - packaging plasmids and transfection reagent, 5 packsComponents: Five kits each containing: 1 vial of packaging plasmids (60 ug) – lyophilized 1 vial of Turbofectin (400 ul) – liquidQuality Control: The reagent components have been successfully tested in transfecting HEK293 cells and producing pseudoviral particles for transduction.Stability: Stable for 1 year from receipt of products under proper storage and handling condition.Storage: Store in 4°C refrigerator upon arrival.Type: Lentiviral Packaging Kits