Learn More
Alstem VP100 Lentiviral Packaging Mix, ready-to-use, 200ul
SDP
Catalog No. NC2734821
Encompass
Lentiviral Packaging Mix is a ready-to-use third generation HIV-based lentiviral packaging system in which the plasmids express the elements required for lentiviral production. To produce high quality lentiviral particles, all you need is a lentiviral expression vector containing your gene of interest. Lentiviral Packaging Mix contains the essential packaging vectors containing the viral structure proteins and the envelope vector to express the Vesicular Stomatitis Virus glycoprotein (VSVG). The lentiviral packaging mix is TAT-independent because it only supports lentiviral expression vector with a chimeric 5' LTR, in which the HIV promoter is replaced with CMV or RSV.
Catalog No. NC2734821 Supplier Alstem Supplier No. VP100
Lentiviral Packaging Mix is a ready-to-use third generation HIV-based lentiviral packaging system in which the plasmids express the elements required for lentiviral production. To produce high quality lentiviral particles, all you need is a lentiviral expression vector containing your gene of interest. Lentiviral Packaging Mix contains the essential packaging vectors containing the viral structure proteins and the envelope vector to express the Vesicular Stomatitis Virus glycoprotein (VSVG). The lentiviral packaging mix is TAT-independent because it only supports lentiviral expression vector with a chimeric 5' LTR, in which the HIV promoter is replaced with CMV or RSV.