Learn More
101 BIO Lentivirus 10X Titer-Up Reagent (Increase virus titer by 5 to 10 times) | 0.1mL
Catalog No. NC0930839
Encompass_Preferred
This virus titer boost is a novel recipe of small molecules designed for increasing / boosting lentivirus packaging efficiency. Lentivirus 10X Titer-Up stably regulates the viral RNA packaging on the transcriptional level, which can enhance production of lentiviral particles for up to 10 fold.
Catalog No. NC0930839 Supplier 101 BIO Supplier No. P906S
Please call customer service for current pricing. 1-800-766-7000
This virus titer boost is a novel recipe of small molecules designed for increasing / boosting lentivirus packaging efficiency. Lentivirus 10X Titer-Up stably regulates the viral RNA packaging on the transcriptional level, which can enhance production of lentiviral particles for up to 10 fold.