Learn More
Gentarget Concentrated lentivirus, 200 μL/vial, 1 x 10^8 IFU/mL, PBS solution, Human Telomerase Reverse Transcriptase (hTERT) expressed, Hygromycin resistance, Enhanced EF1a promoter, RSV promoter
Catalog No. NC3385237
Encompass
Concentrated lentivirus, 200 μL/vial, 1 x 10^8 IFU/mL, PBS solution, Human Telomerase Reverse Transcriptase (hTERT) expressed, Hygromycin resistance, Enhanced EF1a promoter, RSV promoter
Catalog No. NC3385237 Supplier Gentarget Supplier No. LVP1131HYGROPBS
Concentrated lentivirus, 200 μL/vial, 1 x 10^8 IFU/mL, PBS solution, Human Telomerase Reverse Transcriptase (hTERT) expressed, Hygromycin resistance, Enhanced EF1a promoter, RSV promoter