Learn More
TRIBIOSCIENCE INC Lentivirus qPCR Titer Kit, 100 Tests
SDP
Catalog No. NC2887461
Encompass_Preferred
Lentivirus (LV) qPCR Titer Kit incorporates TribioScience FAST technology paradigm and probe qPCR technology for an accurate, one-step, and time-saving qPCR titer determination of lentivirus. The probe is labeled with Fluorescence FAM.
Catalog No. NC2887461 Supplier TRIBIOSCIENCE INC Supplier No. TBS4052100
Lentivirus (LV) qPCR Titer Kit incorporates TribioScience FAST technology paradigm and probe qPCR technology for an accurate, one-step, and time-saving qPCR titer determination of lentivirus. The probe is labeled with Fluorescence FAM.