Learn More
OriGene Lentivirus Stabilizer, 20 mL
SDP
Catalog No. 5000600197
Encompass
Lentivirus stabilizer, preserving the lentiviral infectivity, 20 mLComponents: 1 bottle of sterile Lentivirus Stabilizer solution (20 mL)Quality Control: Lentivirus Stabilizer stabilizes the particles in storage, preserves the infectivity for 1 year.Stability: Stable for 1 year from receipt of products under proper storage and handling condition.Storage: Store at +4°C after receiving vial.
Catalog No. 50-006-00197 Supplier OriGene Supplier No. TR30039L
Lentivirus stabilizer, preserving the lentiviral infectivity, 20 mLComponents: 1 bottle of sterile Lentivirus Stabilizer solution (20 mL)Quality Control: Lentivirus Stabilizer stabilizes the particles in storage, preserves the infectivity for 1 year.Stability: Stable for 1 year from receipt of products under proper storage and handling condition.Storage: Store at +4°C after receiving vial.