Learn More
OriGene Lentivirus Stabilizer, Sterile (5mL)
SDP
Catalog No. NC1484700
Encompass_Preferred
1 bottle of sterile Lentivirus Stabilizer solution (5 mL). Lentivirus Stabilizer stabilizes the particles in storage, preserves the infectivity for 1 year. Store in 4°C refrigerator upon arrival. Stable for 1 year from receipt of products under proper storage and handling condition.
Catalog No. NC1484700 Supplier OriGene Supplier No. TR30039
1 bottle of sterile Lentivirus Stabilizer solution (5 mL). Lentivirus Stabilizer stabilizes the particles in storage, preserves the infectivity for 1 year. Store in 4°C refrigerator upon arrival. Stable for 1 year from receipt of products under proper storage and handling condition.