Learn More
Genscript Corporation Lentivirus Titer p24 ELISA Kit
Supplier:  Genscript Corporation L00938
Encompass
GenScript’s Lentivirus Titer p24 ELISA kit (Cat. No. L00938) is a Sandwich ELISA designed for quantitatively measuring the physical titer of any HIV-1 based lentivirus in cell culture supernatant. For lentivirus purification and production, lentivirus titration is an essential characterization procedure for routine quality control and lot release, as well as for purification process optimization. This kit makes lentivirus titration easy, accurate and reproducible.
Catalog No. NC2137112
 Restricted Item