Learn More
Cellomics Technology LLC LENTIVIRUS TITRATION STRIP (20
SDP
Catalog No. NC1990006
Encompass
Lentivirus titration strip is a quick immunoassay to check lentivirus titers. Imagine how much time and money you could save when you know the lentivirus titers too low to be concentrated before you purify and concentrate the virus.
Catalog No. NC1990006 Supplier Cellomics Technology LLC Supplier No. LVT100120
Lentivirus titration strip is a quick immunoassay to check lentivirus titers. Imagine how much time and money you could save when you know the lentivirus titers too low to be concentrated before you purify and concentrate the virus.