Learn More
Cayman Chemical Lentztrehalose A 10mg
SDP
Supplier:  Cayman Chemical 3773210 MG
Encompass_Preferred
A disaccharide microbial metabolite with diverse biological activities; selectively inhibits M. smegmatis trehalase over porcine kidney trehalase (IC50s = 0.67 and >20 mM, respectively); induces autophagy in MeWo and OVK18 cells at 100 mM; increases survival percentage and decreases tumor proliferation in a Sarcoma 180 murine sarcoma model at 50 mg/kg per day
Catalog No. 50-291-7640